missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinamide N-Methyltransferase/NNMT Antibody (OTI3D8), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72984MFV500
5872.00 SEK valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Nicotinamide N-Methyltransferase/NNMT Monoclonal antibody specifically detects Nicotinamide N-Methyltransferase/NNMT in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
Nicotinamide N-Methyltransferase/NNMT | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI3D8 | |
Flow Cytometry | |
EC 2.1.1.1, nicotinamide N-methyltransferase | |
Full length human recombinant protein of human NNMT(NP_006160) produced in HEK293T cell. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
4837 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction