Läs mer
Nicotinamide Riboside Chloride, TRC
Produktkod. 30011617
Change view
Kvantitet:
250 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011617 1 g -
30009003 2.5 g 2.5g
30010230 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011617 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4124601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23111-00-4
Molekylformel C11 H15 N2 O5 . Cl
Molekylvikt (g/mol) 290.7
Synonym Pyridinium, 3-(aminocarbonyl)-1-β-D-ribofuranosyl-, chloride (1:1), Pyridinium, 3-(aminocarbonyl)-1-β-D-ribofuranosyl-, chloride (9CI), Pyridinium, 3-carbamoyl-1-β-D-ribofuranosyl-, chloride (8CI), N-Ribosylnicotinamide chloride, Nicotinamide riboside chloride
IUPAC-namn 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyridin-1-ium-3-carboxamide;chloride
LEDER [Cl-].NC(=O)c1ccc[n+](c1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O
Specifikationer
Kemiskt namn eller material beta-Nicotinamide riboside chloride
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C11H14N2O5.ClH/c12-10(17)6-2-1-3-13(4-6)11-9(16)8(15)7(5-14)18-11;/h1-4,7-9,11,14-16H,5H2,(H-,12,17);1H/t7-,8-,9-,11-;/m1./s1
Formel vikt 290.0669
Procent renhet >95%