Läs mer
(-)-Nicotine (+)-O,O’-Di-p-toluoyl-D-tartaric Acid, TRC
Produktkod. 30472581
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30472581 25 mg
1 options
Produktkod. 30472581 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN42459025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (-)-Nicotine (+)-O,O’-Di-p-toluoyl-D-tartaric Acid
CAS 68935-25-1
Molekylformel C10H14N2 . (C20H18O8)
Kvantitet 25 mg
Synonym [S-(R*,R*)]-2,3-Bis[(4-methylbenzoyl)oxy]-butanedioic Acid Compd. with (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine, (S)-3-(1-Methyl-2-pyrrolidinyl)-pyridine [S-(R*,R*)]-2,3-Bis[(4-methylbenzoyl)oxy]butanedioate
InChI formel InChI=1S/C20H18O8.C10H14N2/c1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14;1-12-7-3-5-10(12)9-4-2-6-11-8-9/h3-10,15-16H,1-2H3,(H,21,22)(H,23,24);2,4,6,8,10H,3,5,7H2,1H3/t15-,16-;10-/m00/s1
LEDER CN1[C@H](C2=CN=CC=C2)CCC1.O=C(C3=CC=C(C=C3)C)O[C@H](C(O)=O)[C@@H](C(O)=O)OC(C4=CC=C(C=C4)C)=O
IUPAC-namn (S)-3-(1-methylpyrrolidin-2-yl)pyridine (2S,3S)-2,3-bis((4-methylbenzoyl)oxy)succinate
Molekylvikt (g/mol) 548.58
Formel vikt 548.22