Läs mer
Nicotinic Acid-13C1 (~1% unlabeled), TRC
Produktkod. 30471353
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30471353 1 g
1 options
Produktkod. 30471353 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4292541G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nicotinic Acid-13C1
CAS 57987-14-1
Molekylformel C513CH5NO2
Kvantitet 1 g
Synonym 3-Pyridinecarboxylic Acid-13C1, 3-Carboxylpyridine-13C1, 3-Carboxypyridine-13C1 Akotin-13C1, Apelagrin-13C1, Niacin-13C1, Niacor-13C1, Niaspan-13C1, Nicacid-13C1, Pelonin-13C1, Vitamin B3-13C1, Vitamin B5-13C1, Wampocap-13C1, β-Pyridinecarboxylic Acid-13C1
InChI formel InChI=1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)/i6+1
LEDER O[13C](C1=CC=CN=C1)=O
Molekylvikt (g/mol) 124.1
Formel vikt 124.0354