Läs mer
Nicotinic Acid N-Hydroxysuccinimide Ester, TRC
Produktkod. 30479828
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479828 5 mg
30471143 100 mg
30474604 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479828 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4292555MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 78348-28-4
Molekylformel C10 H8 N2 O4
Molekylvikt (g/mol) 220.18
Synonym 3-Pyridinecarboxylic Acid 2,5-Dioxo-1-pyrrolidinyl Ester, N-(3-Pyridinyloyloxy)succinimide, N-(Nicotinoyloxy)succinimide, Succinimidyl Nicotinate
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) pyridine-3-carboxylate
LEDER O=C(ON1C(=O)CCC1=O)c2cccnc2
Specifikationer
Kemiskt namn eller material Nicotinic Acid N-Hydroxysuccinimide Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C10H8N2O4/c13-8-3-4-9(14)12(8)16-10(15)7-2-1-5-11-6-7/h1-2,5-6H,3-4H2
Formel vikt 220.0484
Procent renhet >95%