Läs mer
Nicotinoyl-gamma-aminobutyric Acid, TRC
Produktkod. 30468559
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30468559 5 g
1 options
Produktkod. 30468559 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4312405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nicotinoyl-gamma-aminobutyric Acid
CAS 34562-97-5
Molekylformel C10H12N2O3
Kvantitet 5 g
Synonym 4-Nicotinamidobutyric Acid, 4-[(3-Pyridinylcarbonyl)amino]butanoic Acid, N-Nicotinoyl-γ-aminobutyric Acid, Nicotinoyl-GABA
InChI formel InChI=1S/C10H12N2O3/c13-9(14)4-2-6-12-10(15)8-3-1-5-11-7-8/h1,3,5,7H,2,4,6H2,(H,12,15)(H,13,14)
LEDER O=C(O)CCCNC(C1=CN=CC=C1)=O
Molekylvikt (g/mol) 208.21