missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinuric acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Measurement of NUA levels in urine is used to indicate possible elevated nicotinamide adenine dinucleotide (NAD) catabolism. NUA may be used as a reference (standard curve) in analysis of NAD and NUA catabolism. It is an antiproliferative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Measurement of NUA levels in urine is used to indicate possible elevated nicotinamide adenine dinucleotide (NAD) catabolism. NUA may be used as a reference (standard curve) in analysis of NAD and NUA catabolism. It is an antiproliferative.
Solubility
Soluble in dimethyl sulfoxide, methanol, water, and 1M NH4OH (50 mg/mL).
Notes
Store at -20°C. Incompatible with oxidizing agents.
Measurement of NUA levels in urine is used to indicate possible elevated nicotinamide adenine dinucleotide (NAD) catabolism. NUA may be used as a reference (standard curve) in analysis of NAD and NUA catabolism. It is an antiproliferative.
Solubility
Soluble in dimethyl sulfoxide, methanol, water, and 1M NH4OH (50 mg/mL).
Notes
Store at -20°C. Incompatible with oxidizing agents.
Specifications
Color | White |
Quantity | 25 g |
Solubility Information | Soluble in dimethyl sulfoxide,methanol,water,and 1M NH4OH (50mg/mL). |
Formula Weight | 180.16 |
Percent Purity | ≥98% |
Physical Form | Crystalline Powder |
Chemical Name or Material | Nicotinuric acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'