Läs mer
Nidufexor, MedChemExpress
Produktkod. 16466997
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16466997 5 mg
16460607 50 mg
16455206 100 mg
16444278 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16466997 Leverantör Medchem Express Leverantörsnummer HY109096
MedChemExpress Nidufexor (LMB763) is an orally-available farnesoid X receptor (FXR) agonist for the research of nonalcoholic steatohepatitis (NASH).
Nidufexor (LMB763) is an orally-available farnesoid X receptor (FXR) agonist for the research of nonalcoholic steatohepatitis (NASH).
Specifikationer
Kemiskt namn eller material Nidufexor
CAS 1773489-72-7
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C27H22ClN3O4
Kvantitet 5 mg
Synonym LMB763
Löslighetsinformation DMSO : 83.33 mg/mL (170.78 mM; Need ultrasonic)
LEDER CN1C(C2=CC(Cl)=CC=C2OC3)=C3C(C(N(CC4=CC=CC=C4)CC5=CC=C(C(O)=O)C=C5)=O)=N1
Molekylvikt (g/mol) 487.93
Formel vikt 487.93
Procent renhet 98.51%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Neuroscience-Neuromodulation