Läs mer
Nifedipine EP Impurity C (as E/Z-Mixture), Mikromol™
Produktkod. 30051716
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051716 100 mg
30052595 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051716 Leverantör Mikromol Leverantörsnummer MM0003.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nifedipine EP Impurity C (as E/Z-Mixture)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl (E/Z)-2-(2-Nitrobenzylidene)-3-oxobutanoate
CAS 39562-27-1
Analyt- eller komponentnamn Methyl (E/Z)-2-(2-Nitrobenzylidene)-3-oxobutanoate
Frakt skick Room Temperature
Molekylformel C12 H11 N O5
Certifikat och efterlevnad ISO 17025
Synonym Nifedipine Imp. C (EP), Methyl (E/Z)-2-(2-Nitrobenzylidene)-3-oxobutanoate, Nifedipine Imp. C (EP) as E/Z-Mixture, Nifedipine Impurity C as E/Z-Mixture
LEDER COC(=O)C(=Cc1ccccc1[N+](=O)[O-])C(=O)C
Molekylvikt (g/mol) 249.22
Kvantitet 100 mg
Formel vikt 249.222 g/mol