Läs mer
Nilotinib N-Oxide, TRC
Produktkod. 30470015
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470015 25 mg
30470702 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30470015 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN46531025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246817-85-5
Molekylformel C28 H22 F3 N7 O2
Molekylvikt (g/mol) 545.52
Synonym Benzamide, 4-methyl-N-[3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(1-oxido-3-pyridinyl)-2-pyrimidinyl]amino]-
IUPAC-namn 4-methyl-N-[3-(4-methylimidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(1-oxidopyridin-1-ium-3-yl)pyrimidin-2-yl]amino]benzamide
LEDER Cc1cn(cn1)c2cc(NC(=O)c3ccc(C)c(Nc4nccc(n4)c5ccc[n+]([O-])c5)c3)cc(c2)C(F)(F)F
Specifikationer
Kemiskt namn eller material Nilotinib pyridinyl-N-oxide
Kvantitet 25 mg
InChI formel InChI=1S/C28H22F3N7O2/c1-17-5-6-19(10-25(17)36-27-32-8-7-24(35-27)20-4-3-9-38(40)15-20)26(39)34-22-11-21(28(29,30)31)12-23(13-22)37-14-18(2)33-16-37/h3-16H,1-2H3,(H,34,39)(H,32,35,36)
Formel vikt 545.1787