Läs mer
Nimbanal, TRC
Produktkod. 30005680
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30005680 1 mg
1 options
Produktkod. 30005680 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4762101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nimbanal
CAS 120462-51-3
Molekylformel C29 H34 O8
Kvantitet 1 mg
Synonym (2R,3aR,4aS,5R,5aS,6R,9aR,10S,10aR)-5-(Acetyloxy)-6-formyl-2-(3-furanyl)-3,3a,4a,5,5a,6,9,9a,10,10a-decahydro-1,6,9a,10a-tetramethyl-9-oxo-2H-cyclopenta[b]naphtho[2,3-d]furan-10-acetic Acid Methyl Ester, (4α,5α,6α,7α,15β,17α)-6-(Acetyloxy)-7,15:21,23-diepoxy-4-formyl-4,8-dimethyl-1-oxo-24-nor-12,13-secochola-2,13,20,22-tetraen-12-oic Acid Methyl Ester
InChI formel InChI=1S/C29H34O8/c1-15-18(17-8-10-35-13-17)11-19-23(15)29(5)20(12-22(33)34-6)28(4)21(32)7-9-27(3,14-30)25(28)24(26(29)37-19)36-16(2)31/h7-10,13-14,18-20,24-26H,11-12H2,1-6H3/t18-,19-,20-,24-,25+,26-,27+,28+,29-/m1/s1
LEDER COC(=O)C[C@H]1[C@@]2(C)[C@H](O[C@@H]3C[C@H](C(=C23)C)c4cocc4)[C@H](OC(=O)C)[C@H]5[C@](C)(C=O)C=CC(=O)[C@]15C
Molekylvikt (g/mol) 510.58
Formel vikt 510.2254