Läs mer
Ningetinib, MedChemExpress
Produktkod. 16466807
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16466807 5 mg 5mg
16445076 100 mg 100mg
16424108 10 mg -
16412956 25 mg -
16400467 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16466807 Leverantör Medchem Express Leverantörsnummer HY107145A
MedChemExpress Ningetinib is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and c-Met, VEGFR2 and Axl, respectively.
Ningetinib is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and c-Met, VEGFR2 and Axl, respectively.
Specifikationer
Kemiskt namn eller material Ningetinib
CAS 1394820-69-9
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C31H29FN4O5
Kvantitet 5 mg
Löslighetsinformation DMSO : 16.67 mg/mL (29.95 mM; Need ultrasonic)
LEDER CC1=C(C(NC2=CC(F)=C(OC3=CC=NC4=C3C=CC(OCC(C)(O)C)=C4)C=C2)=O)C(N(C5=CC=CC=C5)N1C)=O
Molekylvikt (g/mol) 556.58
Formel vikt 556.58
Procent renhet 99.79%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease