Läs mer
Nintedanib Esilate, Mikromol™
Produktkod. 18070240
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
18070240 250 mg
1 options
Produktkod. 18070240 Leverantör Mikromol Leverantörsnummer MM3704.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nintedanib Esilate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nintedanib Esilate
CAS 656247-18-6
Analyt- eller komponentnamn Nintedanib Esilate
Frakt skick Room Temperature
Molekylformel C31 H33 N5 O4 . C2 H6 O3 S
Certifikat och efterlevnad ISO 17034
Synonym 1H-Indole-6-carboxylic acid, 2,3-dihydro-3-[[[4-[methyl[(4-methyl-1-piperazinyl)acetyl]amino]phenyl]amino]phenylmethylene]-2-oxo-, methyl ester, (3Z)-, monoethanesulfonate (9CI), Nintedanib esylate, 1H-Indole-6-carboxylic acid, 2,3-dihydro-3-[[[4-[methyl[2-(4-methyl-1-piperazinyl)acetyl]amino]phenyl]amino]phenylmethylene]-2-oxo-, methyl ester, (3Z)-, ethanesulfonate (1:1), Nintedanib ethanesulfonate
InChI formel InChI=1S/C31H33N5O4.C2H6O3S/c1-34-15-17-36(18-16-34)20-27(37)35(2)24-12-10-23(11-13-24)32-29(21-7-5-4-6-8-21)28-25-14-9-22(31(39)40-3)19-26(25)33-30(28)38;1-2-6(3,4)5/h4-14,19,32H,15-18,20H2,1-3H3,(H,33,38);2H2,1H3,(H,3,4,5)/b29-28-;
LEDER CCS(=O)(=O)O.COC(=O)c1ccc2\C(=C(\Nc3ccc(cc3)N(C)C(=O)CN4CCN(C)CC4)/c5ccccc5)\C(=O)Nc2c1
IUPAC-namn ethanesulfonic acid;methyl (3Z)-3-[[4-[methyl-[2-(4-methylpiperazin-1-yl)acetyl]amino]anilino]-phenylmethylidene]-2-oxo-1H-indole-6-carboxylate
Molekylvikt (g/mol) 649.76
Kvantitet 250 mg
Formel vikt 649.257 g/mol