Learn More
Niraparib, MedChemExpress
MedChemExpress Niraparib (MK-4827) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity.
1120.37 SEK - 4497.01 SEK
Specifications
Chemical Name or Material | Niraparib |
---|---|
CAS | 1038915-60-4 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C19H20N4O |
Product Code | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
16465046
![]() |
Medchem Express
HY-10619 |
100 mg |
4497.01 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16460427
![]() |
Medchem Express
HY-10619 |
50 mg |
2894.28 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16426777
![]() |
Medchem Express
HY-10619 |
5 mg |
1120.37 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16404078
![]() |
Medchem Express
HY-10619 |
10 mg |
1602.75 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Niraparib (MK-4827) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity.Spezifikation
Niraparib | |
Off-White | |
C19H20N4O | |
DMSO : 25 mg/mL (78.03 mM; Need ultrasonic) | |
320.39 | |
98.13% | |
Solid |
1038915-60-4 | |
Research | |
MK-4827 | |
NC(C1=CC=CC2=CN(C3=CC=C([C@H]4CNCCC4)C=C3)N=C21)=O | |
320.39 | |
Research | |
Cancer-programmed cell death |
Sicherheit und Handhabung
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.