Learn More
Alfa Aesar™ NIST traceable size standard, polystyrene monodisperse, 15μm, 1% wt%, aqueous suspension in dropper-tipped bottles
Brand:  Alfa Aesar™ J67518.4Y
Additional Details : Weight : 0.00015kg
Product Code. 15484645
  • 4290.46 SEK / 15mL
Estimated Shipment date
from Supplier 06-02-2018
Add to basket
Specifications
Specifications
NIST traceable size standard, polystyrene monodisperse, 15μm
9003-53-6
Liquid
dropper-tipped bottles
15mL
1% wt%, aqueous suspension
1.050g/mL
Odorless