Läs mer
Nitazoxanide, TRC
Produktkod. 30138653
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138653 100 mg
30134177 25 mg
17963995 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138653 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN490100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55981-09-4
Molekylformel C12 H9 N3 O5 S
Molekylvikt (g/mol) 307.28
Synonym Benzamide, 2-(acetyloxy)-N-(5-nitro-2-thiazolyl)-, Alinia, BRN 1225475, Cryptaz, DRG 0242, Daxon, Dexidex, Kidonax, Mitafar, N-(5-Nitro-2-thiazolyl)salicylamide acetate (ester), NSC 697855, NTX, NTZ, Nitacure, Nitacure 500, Nitarid, Nitazode, Nitazox 500, Nitazoxanida, Nitazoxanide, Nixide, Nixoran, Nizonide, PH 5776, Pacovanton, ParaMix, Xerovirin-C
IUPAC-namn [2-[(5-nitro-1,3-thiazol-2-yl)carbamoyl]phenyl] acetate
LEDER CC(=O)Oc1ccccc1C(=O)Nc2ncc(s2)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Nitazoxanide
Kvantitet 100 mg
InChI formel InChI=1S/C12H9N3O5S/c1-7(16)20-9-5-3-2-4-8(9)11(17)14-12-13-6-10(21-12)15(18)19/h2-6H,1H3,(H,13,14,17)
Formel vikt 307.0263