Learn More
Nitinol compression coil spring, wire diameter 1.0mm (0.039in), mean coil diameter 7.0mm (0.28in), Alloy M, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 045951.KN
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nitinol compression coil spring, Alloy M | |
MFCD02091734 | |
Odorless |
Wire diameter 1.0mm (0.039 in.), mean coil diameter 7.0mm (0.27 in.), mean coil diameter 7.0mm (0.28 in.) | |
100 Pc. |