Läs mer
Nitisinone-13C6, TRC
Produktkod. 30180388
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180388 1 mg
30179745 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180388 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4901371MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246815-63-3
Molekylformel C8 13C6 H10 F3 N O5
Molekylvikt (g/mol) 335.18
Synonym Nitisinone 13C6
LEDER [O-][N+](=O)c1cc(ccc1C(=O)[13CH]2[13C](=O)[13CH2][13CH2][13CH2][13C]2=O)C(F)(F)F
Specifikationer
Kemiskt namn eller material Nitisinone-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C14H10F3NO5/c15-14(16,17)7-4-5-8(9(6-7)18(22)23)13(21)12-10(19)2-1-3-11(12)20/h4-6,12H,1-3H2/i1+1,2+1,3+1,10+1,11+1,12+1
Formel vikt 335.0712
Procent renhet >95%