Learn More
Thermo Scientific Chemicals Nitrazine Yellow, indicator grade
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Infrared Spectrum | Authentic |
Formula Weight | 542.35 |
Grade | Indicator |
Packaging | Glass bottle |
UV_Vis | 85% min. |
Visual Transition Interval | from pH 6.0(bright yellow) to pH 7.2(bright blue) |
Color | Brown to Orange |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | Nitrazine yellow |