Läs mer
Nitrendipine Propyl Ester, TRC
Produktkod. 30138366
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138366 100 mg
30006149 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138366 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN490170100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 225785-54-6
Molekylformel C19H22N2O6
Molekylvikt (g/mol) 374.39
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-Methyl 5-Propyl Ester, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid Methyl Propyl Ester;
IUPAC-namn 3-O-methyl 5-O-propyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
LEDER CCCOC(=O)C1=C(NC(=C(C1C2=CC(=CC=C2)[N+](=O)[O-])C(=O)OC)C)C
Specifikationer
Kemiskt namn eller material Nitrendipine Propyl Ester
Kvantitet 100 mg
InChI formel InChI=1S/C19H22N2O6/c1-5-9-27-19(23)16-12(3)20-11(2)15(18(22)26-4)17(16)13-7-6-8-14(10-13)21(24)25/h6-8,10,17,20H,5,9H2,1-4H3
Formel vikt 374.1478