missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric acid concentrate, For 1L standard solution, 1.0 M HNO3 (1.0N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Packaging:
Ampoule
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 100°C |
Density | 1.200 g/cm3 |
Packaging | Ampoule |
Quantity | 6 x 1 Ea. |
Linear Formula | HNO3 |
Physical Form | Concentrate |
UN Number | UN2031 |
Formula Weight | 63.01 |
Chemical Name or Material | Nitric acid concentrate, For 1L standard solution, 1.0 M HNO3 (1.0N) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'