missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Nitric acid concentrate, For 1L standard solution, 0.1 M HNO3 (0.1N), Honeywell Fluka™

for 1L standard solution, 1.0 M HNO3 (1.0N)

449.30 SEK - 2579.45 SEK

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.012
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES [N+](=O)(O)[O-]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15683650 View Documents Honeywell Fluka™
32042-1EA
1 ea. Ampoule 494.52 SEK
Each View alternative products
Save Up to 
»
EU Stock 14
Add to basket
 
15693650 SDS View Documents Honeywell Fluka™
32042-6X1EA
6 x 1 ea. Ampoule 2579.45 SEK
Pack of 6 View alternative products
Save Up to 
»
EU Stock 19
Add to basket
 
15605270 View Documents Honeywell Fluka™
38274-6X1EA
6 x 1 ea. Ampoule 2325.59 SEK
Pack of 6 View alternative products
Save Up to 
»
Call for latest availability.
+46 (0)31 352 32 30

Add to basket
 
15695260 View Documents Honeywell Fluka™
38274-1EA
1 pc. Ampoule 449.30 SEK
Each View alternative products
Save Up to 
»
EU Stock 4
Add to basket
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7697-37-2
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
[N+](=O)(O)[O-]

Specifications

Nitric Acid Concentrate
7697-37-2
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
[N+](=O)(O)[O-]
63.012
CHEBI:48107
Ampoule
1 ea.
HNO3
2031
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
63.01g/mol