Läs mer
Nitrilotris(methylene)triphosphonic Acid, TRC
Produktkod. 30476401
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30476401 1 g
1 options
Produktkod. 30476401 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4902001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nitrilotris(methylene)triphosphonic Acid
CAS 6419-19-8
Molekylformel C3 H12 N O9 P3
Kvantitet 1 g
Synonym P,P’,P’’-[nitrilotris(methylene)]trisphosphonic Acid, 1,1,1-Nitrilotris(methylphosphonic Acid), ATMP, Aminotri(methylenephosphonic Acid), Briquest 301, NTPH, Nitrilo-
InChI formel InChI=1S/C3H12NO9P3/c5-14(6,7)1-4(2-15(8,9)10)3-16(11,12)13/h1-3H2,(H2,5,6,7)(H2,8,9,10)(H2,11,12,13)
LEDER OP(=O)(O)CN(CP(=O)(O)O)CP(=O)(O)O
IUPAC-namn [bis(phosphonomethyl)amino]methylphosphonic acid
Molekylvikt (g/mol) 299.05
Formel vikt 298.9725