Läs mer
Nitrite Standard: NO2- @ 10000 μg/mL in H2O, VHG™
Produktkod. 17957424
Change view
Kvantitet:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17957424 500 ml
17949704 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17957424 Leverantör VHG Labs Leverantörsnummer VHGI1PNO2500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Nitrite Standard: NO2- @ 10000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nitrite
CAS 14797-65-0
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnamn Nitrite
Frakt skick Cold 2°c to 8°C
Molekylformel (N O2)-
Certifikat och efterlevnad ISO 17034
Synonym NO2−
LEDER [O-]N=O
IUPAC-namn Nitrite
Lösningsmedel eller matris Water
Molekylvikt (g/mol) 46.01
Kvantitet 500 ml
Formel vikt 46.01 g/mol