Learn More
Ansell BioClean™ Maxima BLLS Sterile Latex Cleanroom Gloves
Catalog Number 12745785
Click to view available options
Size: 7
This item is not returnable. View return policy
This item is not returnable. View return policy
Sterile latex cleanroom glove provides extra coverage and protection to the upper arm. Recommended for Class 10/ISO 4 cleanroom environments.
Sterile latex cleanroom glove providing extra coverage and protection to the upper arm. Recommended for Class 10/ISO 4 cleanroom environments.
  • Natural color
  • Hand-specific, anatomically shaped
  • Textured surface for enhanced grip
  • Beaded cuff for stability on the arm
  • Sterility Assurance Level 10-6