Learn More
Ansell BioClean™ Maxima BLLS Sterile Latex Cleanroom Gloves
Click to view available options
Size: 8
Description
Sterile latex cleanroom glove providing extra coverage and protection to the upper arm. Recommended for Class 10/ISO 4 cleanroom environments.
- Natural color
- Hand-specific, anatomically shaped
- Textured surface for enhanced grip
- Beaded cuff for stability on the arm
- Sterility Assurance Level 10-6