Läs mer
Nitromifene Citrate (E/Z mixture), TRC
Produktkod. 30472312
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30472312 10 mg
1 options
Produktkod. 30472312 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN49670010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nitromifene Citrate (E/Z mixture)
CAS 5863-35-4
Molekylformel C33H36N2O11
Kvantitet 10 mg
Synonym 1-[2-[4-[1-(4-Methoxyphenyl)-2-nitro-2-phenylethenyl]phenoxy]ethyl]-pyrrolidine 2-Hydroxy-1,2,3-propanetricarboxylate, 1-[2-[p-[α-(p-Methoxyphenyl)-β-nitrostyryl]phenoxy]ethyl]-pyrrolidine Citrate, 1-[2-(p-[a-(p-Methoxyphenyl)-β-nitrostyryl]-phenoxy)ethyl]pyrrolidine Monocitrate, 1-[2-[4-[2-(4-Methoxyphenyl)-1-nitro-2-phenylethenyl]phenoxy]ethyl]pyrrolidine Monocitrate, 1-[2-[p-[α-(p-Methoxyphenyl)-β-nitrostyryl]phenoxy]ethyl]pyrrolidine Monocitrate, 1-[2-[p-[α-(p-Methoxyphenyl)-β-nitrostyryl]phenoxy]ethyl]pyrrollidine Monocitrate, CI 628, CN 55945-27, NSC 110407, Parke Davis CI-628
InChI formel InChI=1S/C27H28N2O4.C6H8O7/c1-32-24-13-9-21(10-14-24)26(27(29(30)31)23-7-3-2-4-8-23)22-11-15-25(16-12-22)33-20-19-28-17-5-6-18-28;7-3(8)1-6(13,5(11)12)2-4(9)10/h2-4,7-16H,5-6,17-20H2,1H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b27-26-;
LEDER COC(C=C1)=CC=C1/C(C2=CC=C(OCCN3CCCC3)C=C2)=C([N+]([O-])=O)/C4=CC=CC=C4.OC(CC(O)=O)(CC(O)=O)C(O)=O
IUPAC-namn 2-hydroxypropane-1,2,3-tricarboxylic acid;1-[2-[4-[(Z)-1-(4-methoxyphenyl)-2-nitro-2-phenylethenyl]phenoxy]ethyl]pyrrolidine
Molekylvikt (g/mol) 636.65
Formel vikt 636.2319