missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Nitrotough™ N110 Series Light Duty Oil Repellent Gloves

Catalog Number 15620934
Click to view available options
Size:
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed specifically for use in moderately oily manufacturing and maintenance environments. Ansell Edmont™ Nitrotough™ N110 Series Light Duty Oil Repellent Gloves have high degree of flexibility and durability with optimum dexterity.

  • Mechanical protection gloves
  • Light duty oil repellent
  • Ideal alternative to light leather gloves
  • Fine and stretchable stitches for good comfort
  • Used in environments where protection combined with abrasion resistance are essential

Applications:

  • Assembly and finishing
  • Engineering applications
  • Screwing and bolting
Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2006
Color Black, White
Exterior Finish Palm Coated
Material Nitrile
Product Type Oil repellent gloves
Size 8
Style Construction: Knitted
Glove Cuff Style Knitwrist
Packaging Type 12 pairs in a bag, 12 bags in a carton
Series N110
Length (Metric) 230 to 260 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'