Läs mer
Nizatidine Amide (>90%), TRC
Produktkod. 30465239
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30465239 100 mg
1 options
Produktkod. 30465239 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN598520100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nizatidine Amide
CAS 188666-11-7
Molekylformel C11H18N4O3S2
Kvantitet 100 mg
Synonym N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-2-nitroacetamide, Nizatidine EP Impurity E
InChI formel InChI=1S/C11H18N4O3S2/c1-14(2)6-11-13-9(8-20-11)7-19-4-3-12-10(16)5-15(17)18/h8H,3-7H2,1-2H3,(H,12,16)
LEDER CN(C)Cc1nc(CSCCNC(=O)C[N+](=O)[O-])cs1
Molekylvikt (g/mol) 318.42