Läs mer
Nizatidine EP Impurity A, Mikromol™
Produktkod. 30051857
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051857 25 mg
30051087 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051857 Leverantör Mikromol Leverantörsnummer MM0220.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nizatidine EP Impurity A
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N,N'-Dimethyl-2-nitroethene-1,1-diamine
CAS 54252-45-8
Analyt- eller komponentnamn N,N'-Dimethyl-2-nitroethene-1,1-diamine
Frakt skick Room Temperature
Molekylformel C4 H9 N3 O2
Certifikat och efterlevnad ISO 9001
Synonym N,N'-Dimethyl-2-nitroethene-1,1-diamine, Nizatidine Imp. A (EP), Nizatidine Impurity A
InChI formel InChI=1S/C4H9N3O2/c1-5-4(6-2)3-7(8)9/h3,5-6H,1-2H3
LEDER CNC(=C[N+](=O)[O-])NC
IUPAC-namn 1-N,1-N'-dimethyl-2-nitroethene-1,1-diamine
Molekylvikt (g/mol) 131.13
Kvantitet 25 mg
Formel vikt 131.0695 g/mol