Learn More
NKAIN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553859
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SKDTDLMTFN ISVHRSWWRE HGPGCVRRVL PPSAHGMMDD YTYVSVTGCI VDFQYLEV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.
NKAIN3 Polyclonal antibody specifically detects NKAIN3 in Human samples. It is validated for Immunohistochemistry.Specifications
NKAIN3 | |
Polyclonal | |
Unconjugated | |
NKAIN3 | |
FAM77D; family with sequence similarity 77, member D; Na(+)/K(+)-transporting ATPase subunit beta-1-interacting protein 3; Na+/K+ transporting ATPase interacting 3; NKAIN3; sodium/potassium-transporting ATPase subunit beta-1-interacting protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286183 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N8D7 | |
NKAIN3 | |
Recombinant protein corresponding to Human NKAIN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only