missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKCC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18970-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The sodium-potassium-chloride cotransporter isoform 2 is kidney-specific and is found on the apical membrane of the thick ascending limb of Henles loop and the macula densa. It accounts for most of the NaCl resorption with the stoichiometry of 1Na:1K:2Cl and is sensitive to such diuretics as furosemide and bumetanide. Some Bartter-like syndromes result from defects in this gene.Specifications
NKCC2 | |
Polyclonal | |
Unconjugated | |
SLC12A1 | |
BSC1, NKCC2, SLC12A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20495, 25065, 6557 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P55014, P55016, Q13621 | |
SLC12A1 | |
NKCC2 Fusion Protein Ag5325 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |