Learn More
NKG2AB6 Monoclonal Antibody (16a11), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1240.00 SEK - 3205.00 SEK
Specifications
Antigen | NKG2AB6 |
---|---|
Clone | 16a11 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15559306
|
Affymetrix eBioscience
46-5897-82 |
100 μg |
3205.00 SEK
100µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15549306
|
Affymetrix eBioscience
46-5897-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 16a11 monoclonal antibody reacts with the mouse NKG2A^B6. NKG2 molecules belong to a C-type lectin-like family of cell surface receptors expressed by mouse NK and NKT cell lineages. NKG2 molecules form heterodimeric complexes with CD95 and are responsible for recognition of non-classical MHC class I antigen Qa-1. This antibody reacts with the C57Bl/6 mouse strain, but does not react with the BALB/c or 129 mouse strains. When co-staining with NKG2A/C/E clone 20d5, it is important to stain for NKG2A^B6 first then subsequently with NKG2A/C/E, as steric hindrance has been observed if co-stained concurrently. Applications Reported: This 16a11 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 16a11 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative.
NKG2AB6 Monoclonal antibody specifically detects NKG2AB6 in Mouse samples. It is validated for Flow Cytometry.Specifications
NKG2AB6 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
16641 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
16a11 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Mouse | |
Klrc1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Klrc1 |
For Research Use Only.