missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Nkx2.2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540526
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Peptide sequence: MSLTNTKTGF SVKDILDLPD TNDEEGSVAE GPEEENEGPE PAKRAGPLGQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 100%; Sheep: 100%.
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
Specifikationer
| Nkx2.2 | |
| Polyclonal | |
| Unconjugated | |
| NKX2-2 | |
| Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18088, 4821 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95096, P42586 | |
| NKX2-2 | |
| Synthetic peptide directed towards the N-terminal of human NKX2-2 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering