missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Nkx2.2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582096
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: SLTNTKTGFS VKDILDLPDT NDEEGSVAEG PEEENEGPEP AKRAGPLGQG ALDAVQSLPL KNPFYDSSDN PYTRWLASTE GLQYSLHGLA AGAPPQDSSS KSPEPSADES PDNDKETP Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 96%.
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
Specifikationer
| Nkx2.2 | |
| Polyclonal | |
| Unconjugated | |
| NKX2-2 | |
| Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4821 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95096 | |
| NKX2-2 | |
| Recombinant protein corresponding to Human NKX2-2. Recombinant protein control fragment (Product #RP-100713). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering