Learn More
Nkx3.2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555657
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PKPRKKRSRA AFSHAQVFEL ERRFNHQRYL SGPERADLAA SLKLTETQVK IWFQNRRYKT KRRQMAADLL ASAPAAKKVA VKVLVRDDQR QYLPGEVLRP PSLLPLQPSY YYPYY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Nkx3.2 Polyclonal antibody specifically detects Nkx3.2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Nkx3.2 | |
Polyclonal | |
Unconjugated | |
NKX3-2 | |
bagpipe homeobox gene 1 homolog; bagpipe homeobox protein homolog 1; BAPX1; homeobox protein NK-3 homolog B; Homeobox protein Nkx-3.2; homeodomain protein Nkx-3.2; NK3 homeobox 2; NKX3.2; Nkx-3.2; NKX3-2; NKX3B; SMMD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
579 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P78367 | |
NKX3-2 | |
Recombinant protein corresponding to Human Nkx3.2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only