missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ [Nle13, Glu14] Motilin, Human, Porcine

For cellular and molecular biology applications

1416.00 SEK - 2300.00 SEK

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 2682.10
Product Type [Nle13, Glu14] Motilin, human, porcine
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15497559
SDS View Documents Promotion Details
Thermo Scientific™
J66270.!D
0.5 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15407569
SDS View Documents Promotion Details
Thermo Scientific™
J66270.M^
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

HPLC
Solid
[Nle13, Glu14] Motilin, human, porcine
Moisture sensitive
-30°C to -10°C
2682.10
RUO - research use only
Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Nle-Glu-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only