missing translation for 'onlineSavingsMsg'
Learn More

NLGN4Y Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13489-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881724

  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody can recognize both of NLGN4Y and NLGN4X at 72 kDa mature form.

Neuroligins, such as NLGN4Y, are cell adhesion molecules present at the postsynaptic side of the synapse and may be essential for the formation of functional synapses (Jamain et al., 2003 ).
Specifications

Specifications

NLGN4Y
Polyclonal
Unconjugated
NLGN4Y
KIAA0951, neuroligin 4, Y linked, Neuroligin Y, NLGN4Y
Rabbit
Antigen Affinity Chromatography
RUO
22829, 57502
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N0W4, Q8NFZ3
NLGN4X, NLGN4Y
NLGN4Y Fusion Protein Ag4311
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers