Läs mer
Schmidlin™ NM Plus Hydrogen Generator
Produktkod. 17708919
Encompass_Preferred
Change view
Flödeshastighet:
100 ml/min
1000 ml/min
250 ml/min
500 ml/min
Wattal:
100 W
165 W
220 W
385 W
Förpackningsstorlek:
Varje
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Flödeshastighet Wattal unitSize
17708919 100 ml/min 100 W Varje
17747384 250 ml/min 165 W Varje
17728919 500 ml/min 220 W Varje
17718919 1000 ml/min 385 W Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 17708919 Leverantör Schmidlin™ Leverantörsnummer DBPNM100EU
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This generator produces a continuous supply of high purity, dry hydrogen.
  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Pressure: 11 barg (160 psig)
  • Unique permeation membrane drying system
  • Cold dual dynamic regeneration dryer
  • USB connectivity
  • Easy to install with complete 'plug and play' functionality
  • Ideal for all GC detector applications
  • Meets and exceeds requirements for the most demanding GC applications
  • Superior hydrogen production with reliable long life cell
  • Minimal maintenance; no desiccant cartridges to change
  • PC monitoring for maintenance, diagnostics and remote connection
Specifikationer
Djup (metriskt) 37 cm
Flödeshastighet 100 ml/min
Höjd (metrisk) 50.7 cm
Max. Omgivningstemperatur 35°C
Utloppstryck 11 bar
Bredd (engelska) 9 in.
Bredd (metrisk) 23 cm
Anslutningar Outlet: 1/8 in.
Elektriska krav 110/120 V at 60 Hz, 220/240 V at 50 Hz
Kväve renhet >99.99996%
Wattal 100 W