Läs mer
NMDA-IN-1, MedChemExpress
Produktkod. 16448167
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16448167 5 mg
16405458 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16448167 Leverantör Medchem Express Leverantörsnummer HY12962
MedChemExpress NMDA-IN-1 is a potent and NR2B-selective NMDA antagonist with Ki of 0.85 nM; NR2B Ca2+ influx IC50 is 9.7 nM; no activities on NR2A, NR2C, NR2D, hERG-channel and α1-adrenergic receptor.
NMDA-IN-1 is a potent and NR2B-selective NMDA antagonist with Ki of 0.85 nM
Specifikationer
Kemiskt namn eller material NMDA-IN-1
CAS 700878-19-9
Färg Gray
Anteckningar om renhetsgrad Research
Molekylformel C20H23ClFN3O
Kvantitet 5 mg
Löslighetsinformation DMSO : 12.5 mg/mL (33.26 mM; ultrasonic and warming and heat to 60°C)
LEDER OC1=CC=C2N=C(CN3CCC(CC4=CC=CC=C4F)CC3)NC2=C1.[H]Cl
Molekylvikt (g/mol) 375.87
Formel vikt 375.87
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neurodegeneration