Läs mer
Abnova™ NME2 Recombinant Protein
Beskrivning
Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by NME1) and 'B' (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants encoding the same isoform have been found for this gene. Co-transcription of this gene and the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) which encode a fusion protein comprised of sequence sharing identity with each individual gene product.
- Human NME2 full-length ORF ( AAH02476, 1 a.a. - 152 a.a.) recombinant protein with GST-tag at N-terminal
- Gene description: protein (NM23B) expressed in non-metastatic cells 2
- Theoretical molecular weight: 42.46kDa
- Preparation method: in vitro wheat germ expression system
- Purification: glutathione sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer
Sequence: MANLERTFIAIKPDGVQRGLVGEIIKRFEQKGFRLVAMKFLRASEEHLKQHYIDLKDRPFFPGLVKYMNSGPVVAMVWEGLNVVKTGRVMLGETNPADSKPGTIRGDFCIQVGRNIIHGSDSVKSAEKEISLWFKPEELVDYKSCAHDWVYE
Best use within three months from the date of receipt.
ELISA, western blotting (recombinant protein), antibody production, protein array
Specifikationer
Specifikationer
| Tillträdesnummer | AAH02476 |
| För användning med (applikation) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulering | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| Gen-ID (Entrez) | 4831 |
| Molekylvikt (g/mol) | 42.46 |
| Namn | NME2 (Human) Recombinant Protein (P01) |
| pH-intervall | 8 |
| Beredningsmetod | In vitro wheat germ expression system |
| Reningsmetod | Glutathione Sepharose 4 Fast Flow |
| Kvalitetskontrolltestning | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Visa mer |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.