missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NME7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550244
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total NME7 protein.
NME7 Polyclonal antibody specifically detects NME7 in Human samples. It is validated for Western Blot.Specifications
NME7 | |
Polyclonal | |
Unconjugated | |
NME7 | |
CFAP67; cilia and flagella associated protein 67; D530024H21Rik; MN23H7; NDK 7; NDK7; NDP kinase 7; nm23-H7; Nm23-M7; Nm23-r7; NME/NM23 family member 7; Nme7; non-metastatic cells 7, protein expressed in (nucleoside-diphosphate kinase); nucleoside diphosphate kinase 7; nucleoside diphosphate kinase 7-like protein; nucleoside-diphosphate kinase; nucleoside-diphosphate kinase 7; nucleoside-diphosphate kinase 7 isoform a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29922 | |
-20°C | |
Liquid |
Western Blot | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9Y5B8 | |
NME7 | |
Fusion protein corresponding to residues near the C terminal of human NME/NM23 family member 7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |