Learn More
NMNAT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563436
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VDPWESEQAQ WMETVKVLRH HHSKLLRSPP QMEGPDHGKA LFSTPAAVPE LKLLCGADVL KTFQTPNLWK DAHIQEIVEK FGLVCV Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.
NMNAT3 Polyclonal antibody specifically detects NMNAT3 in Human samples. It is validated for Immunohistochemistry.Specifications
NMNAT3 | |
Polyclonal | |
Unconjugated | |
NMNAT3 | |
4933408N02Rik; FKSG76; naMN adenylyltransferase 1; naMN adenylyltransferase 3; nicotinamide mononucleotide adenylyltransferase 3; nicotinamide nucleotide adenylyltransferase 3; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 3; nicotinamide-nucleotide adenylyltransferase 3; nicotinate-nucleotide adenylyltransferase 1; nicotinate-nucleotide adenylyltransferase 3; NMN adenylyltransferase 3; NMN/NaMN adenylyltransferase 3; NMNAT3; PNAT3; PNAT-3; pyridine nucleotide adenylyltransferase 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
349565 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96T66 | |
NMNAT3 | |
Recombinant protein corresponding to Human NMNAT3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only