Läs mer
NMS-P118, MedChemExpress
Produktkod. 16490306
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
10mg
25mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16490306 2 mg -
16490118 5 mg -
16483616 25 mg 25mg
16477556 100 mg 100mg
16443667 50 mg -
16427658 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16490306 Leverantör Medchem Express Leverantörsnummer HY18954
MedChemExpress NMS-P118 is a potent, orally available, and highly selective PARP-1 Inhibitor for cancer therapy.
NMS-P118 is a potent, orally available, and highly selective PARP-1 Inhibitor for cancer therapy.
Specifikationer
Kemiskt namn eller material NMS-P118
CAS 1262417-51-5
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C20H24F3N3O2
Kvantitet 2 mg
Löslighetsinformation DMSO : 16 mg/mL (40.46 mM; Need ultrasonic and warming)
LEDER FC(CC1)(F)CCC1N(CC2)CCC2N(C3=O)CC4=C3C(C(N)=O)=CC(F)=C4
Molekylvikt (g/mol) 395.42
Formel vikt 395.42
Procent renhet 99.8%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death