Learn More
ZNF330 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566009
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: RKKAENRRERE KQLRASRSTI DLAKHPCNAS MECDKCQRRQ KNRAFCYFCN SVQKLPICAQ CGKTKCMMKS SDCVIKHAGV YSTGLAMVGA ICDFCEAWVC HGRKCLSTHA CACPLTDAEC VECER Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
ZNF330 Polyclonal antibody specifically detects ZNF330 in Human samples. It is validated for Immunocytochemistry.Specifications
ZNF330 | |
Polyclonal | |
Unconjugated | |
ZNF330 | |
BC008086; HSA6591; NOA 36; Noa36; Nucleolar autoantigen 36; nucleolar cysteine-rich protein; Zfp330; zinc finger 330-like protein; zinc finger autoantigen 330; zinc finger protein 330; zinc finger protein 330-like protein; Znf330 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27309 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y3S2 | |
ZNF330 | |
Recombinant Human NOA36 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only