missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
NOD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-76286-0.025mg
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NOD1 Polyclonal specifically detects NOD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifikationer
| NOD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| NOD1 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy end of human NOD1. The immunogen is located within the last 50 amino acids of NOD1. Amino Acid Squence: NLIKEPEEAKVYEDEKR | |
| 0.025 mg | |
| Apoptosis, Asthma, Autophagy, Immunology | |
| 10392 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000 | |
| CARD4NLR family, CARD domain containing 1, caspase recruitment domain family, member 4, Caspase recruitment domain-containing protein 4, CLR7.1, NLRC1, nucleotide-binding oligomerization domain containing 1, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 1, nucleotide-binding oligomerization domain-containing protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering