Läs mer
Nogalamycin, TRC
Produktkod. 30421428
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421428 1 mg
30426440 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30421428 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN8885031MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nogalamycin
CAS 1404-15-5
Molekylformel C39 H49 N O16
Kvantitet 1 mg
Synonym Antibiotic 205t3, NSC 70845, U 15167, [2R-(2α,3β,4α,5β,6α,11β,13α,14α)]-11-[(6-Deoxy-3-C-methyl-2,3,4-tri-O-methyl-α-L-mannopyranosyl)oxy]-4-(dimethylamino)-3,4,5,6,9,11,12,13,14,16-decahydro-3,5,8,10,13-pentahydroxy-6,13-dimethyl-9,16-dioxo-methyl Ester 2,6-Epoxy-2H-naphthaceno[1,2-b]oxocin-14-carboxylic Acid
InChI formel InChI=1S/C39H49NO16/c1-14-32(49-7)39(4,52-10)33(50-8)36(53-14)54-19-13-37(2,48)24(34(47)51-9)15-11-16-21(27(43)20(15)19)28(44)22-18(41)12-17-30(23(22)26(16)42)55-35-29(45)25(40(5)6)31(46)38(17,3)56-35/h11-12,14,19,24-25,29,31-33,35-36,41,43,45-46,48H,13H2
LEDER CO[C@H]1[C@H](C)O[C@@H](O[C@H]2C[C@](C)(O)[C@H](C(=O)OC)c3cc4C(=O)c5c6O[C@@H]7O[C@@](C)([C@H](O)[C@H]([C@@H]7O)N(C)C)c6cc(O)c5C(=O)c4c(O)c23)[C@H](OC)[C@]1(C)OC
Molekylvikt (g/mol) 787.803
Formel vikt 787.305