missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568254
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Human, Mouse, and Rat samples.
NOL11 is a ribosome biogenesis factor. It may be required for both optimal rDNA transcription and small subunit (SSU) pre-rRNA processing at sites A′, A0, 1 and 2b.Specifications
NOL11 | |
Polyclonal | |
Unconjugated | |
NOL11 | |
NOL11 | |
Synthesized peptide derived from human NOL11. | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9H8H0, Q8BJW5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25926, 688885, 68979, 718803 | |
-20°C | |
Liquid |