Läs mer
Greiner Bio-One Non-treated 384-well Microplates
Färg:
Black
Kvantitet:
40/Cs.
Lock:
Without Lid
Total volym (metrisk) Tja:
131 μL
Beskrivning
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Quadrupled the number of wells (from 96)
- Well-to-well spacing: 4.5 mm
- Rounded square well profile, providing flexible working volume of 10 μl to 130 μl and reduced wicking and bubbling
Specifications
Specifications
| Certifikat och efterlevnad | DIN ISO 9001 |
| Formatera | Standard |
| Lock | Without Lid |
| Array | 16 x 24 |
| Bindningstyp | Medium Binding |
| Färg | Black |
| Material | Polystyrene |
| Antal Wells | 384 |
| Sterilitet | Non-sterile |
| Ytbehandling | Non-treated |
| Show More |