Learn More
Nonafluor-1-butansulfonsyra (50 μg/ml i acetonitril), TRC
Specifications
Specifications
| Kemiskt namn eller material | Perfluorobutanesulfonic acid |
| CAS | 375-73-5 |
| Molekylformel | C4 H F9 O3 S |
| Kvantitet | 5 mL |
| Synonym | 1-Butanesulfonic acid, nonafluoro- (6CI,7CI,8CI), 1,1,2,2,3,3,4,4,4-Nonafluoro-1-butanesulfonic acid, 1-Perfluorobutanesulfonic acid, Eftop FBSA, Nonafluoro-1-butanesulfonic acid, Nonafluorobutanesulfonic acid, Perfluorobutanesulfonic acid, PFBS |
| InChI formel | InChI=1S/C4HF9O3S/c5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16/h(H,14,15,16) |
| LEDER | OS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC-namn | 1,1,2,2,3,3,4,4,4-nonafluorobutan-1-sulfonsyra |
| Molekylvikt (g/mol) | 300.1 |
| Formel vikt | 299.9503 |
| Show More |